Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308147.1 | NP_001295076.1 | 10 - 11 | 1542 | 114 |
XM_011536624.2 | XP_011534926.1 | 10 - 11 | 1370 | 114 | |
XM_011536625.2 | XP_011534927.1 | 10 - 11 | 1330 | 114 | |
XM_011536627.2 | XP_011534929.1 | 10 - 11 | 1635 | 114 | |
XM_011536628.2 | XP_011534930.1 | 10 - 11 | 1821 | 114 | |
XM_011536629.2 | XP_011534931.1 | 10 - 11 | 1322 | 114 | |
XM_017021154.1 | XP_016876643.1 | 10 - 11 | 1326 | 114 | |
XM_017021155.1 | XP_016876644.1 | 10 - 11 | 1325 | 114 | |
XM_017021156.1 | XP_016876645.1 | 10 - 11 | 1393 | 114 | |
XM_017021157.1 | XP_016876646.1 | 10 - 11 | 1642 | 114 | |
XM_017021158.1 | XP_016876647.1 | 8 - 9 | 828 | 114 | |
GenBank mRNA | AK074169.1 | - | 5 - 6 | 849 | 114 |
BC129953.1 | - | 10 - 11 | 1353 | 114 | |
BX248779.1 | - | 8 - 9 | 1346 | 114 |