Assay Design
Gene Transcripts
- Gene Symbol
- ZFYVE26
- Entrez Gene ID
-
23503
- Gene Name
- zinc finger FYVE-type containing 26
- Gene Aliases
-
FYVE-CENT, SPG15
- Chromosome Location
-
Chr.14: 67728892 - 67817293 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC008927.2 |
- |
6 - 7 |
910 |
90 |
View Full Product Details