Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252049.1 | NP_001238978.1 | 7 - 8 | 878 | 81 |
NM_001252052.1 | NP_001238981.1 | 6 - 7 | 773 | 81 | |
XM_011535868.2 | XP_011534170.1 | 7 - 8 | 1079 | 81 | |
GenBank mRNA | AL539512.3 | - | 6 - 7 | 745 | 81 |
BC007501.2 | - | 7 - 8 | 820 | 81 | |
D13892.1 | - | 7 - 8 | 821 | 81 | |
D25546.1 | - | 7 - 8 | 744 | 81 | |
M93008.1 | - | 7 - 8 | 708 | 81 |