Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004799.3 | NP_004790.2 | 2 - 3 | 418 | 82 |
NM_007324.3 | NP_015563.2 | 2 - 3 | 418 | 82 | |
XM_011542437.2 | XP_011540739.1 | 2 - 3 | 289 | 82 | |
XM_017002845.1 | XP_016858334.1 | 2 - 3 | 511 | 82 | |
GenBank mRNA | AF104304.1 | - | 1 - 2 | 97 | 82 |
AF130419.1 | - | 2 - 3 | 138 | 82 | |
AF130420.1 | - | 2 - 3 | 138 | 82 | |
DR005493.1 | - | 2 - 3 | 420 | 82 |