Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040151.1 | NP_001035241.1 | 6 - 7 | 1080 | 95 |
NM_018400.3 | NP_060870.1 | 5 - 6 | 1479 | 95 | |
GenBank mRNA | AB032984.1 | - | 6 - 7 | 720 | 95 |
AJ243396.2 | - | 5 - 6 | 1048 | 95 | |
AL136589.1 | - | 5 - 6 | 1476 | 95 | |
BC117282.1 | - | 5 - 6 | 766 | 95 | |
BC126265.1 | - | 5 - 6 | 766 | 95 |