Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005173.3 | NP_005164.2 | 8 - 9 | 1268 | 53 |
NM_174953.2 | NP_777613.1 | 8 - 9 | 1268 | 53 | |
NM_174954.2 | NP_777614.1 | 8 - 9 | 1268 | 53 | |
NM_174955.2 | NP_777615.1 | 8 - 9 | 1268 | 53 | |
NM_174956.2 | NP_777616.1 | 8 - 9 | 1268 | 53 | |
NM_174957.2 | NP_777617.1 | 8 - 9 | 1268 | 53 | |
NM_174958.2 | NP_777618.1 | 8 - 9 | 1268 | 53 | |
XM_011523881.2 | XP_011522183.1 | 8 - 9 | 1268 | 53 | |
XM_011523882.2 | XP_011522184.1 | 8 - 9 | 1268 | 53 | |
XM_011523884.2 | XP_011522186.1 | 8 - 9 | 1268 | 53 | |
XM_011523885.1 | XP_011522187.1 | 8 - 9 | 1268 | 53 | |
XM_011523888.2 | XP_011522190.1 | 8 - 9 | 1268 | 53 | |
XM_011523889.1 | XP_011522191.1 | 8 - 9 | 1268 | 53 | |
XM_011523892.2 | XP_011522194.1 | 8 - 9 | 1268 | 53 | |
XM_017024692.1 | XP_016880181.1 | 8 - 9 | 1268 | 53 | |
XM_017024693.1 | XP_016880182.1 | 6 - 7 | 894 | 53 | |
GenBank mRNA | AF068220.1 | - | 8 - 9 | 1219 | 53 |
AK292716.1 | - | 8 - 9 | 1268 | 53 | |
BC035729.1 | - | 8 - 9 | 1223 | 53 | |
Z69881.1 | - | 8 - 9 | 1101 | 53 |