Assay Design



Gene Transcripts

Gene Symbol
COX4I2
Entrez Gene ID
84701
Gene Name
cytochrome c oxidase subunit 4I2
Gene Aliases
COX4, COX4-2, COX4B, COX4L2, COXIV-2, dJ857M17.2
Chromosome Location
Chr.20: 31637856 - 31645009 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032609.2 NP_115998.2 1 - 2 80 104
XM_005260581.3 XP_005260638.1 1 - 2 112 104
GenBank mRNA AF257180.1 - 1 - 2 80 104

View Full Product Details