Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152550.3 | NP_689763.3 | 5 - 6 | 1297 | 117 |
XM_005268380.2 | XP_005268437.1 | 5 - 6 | 1293 | 117 | |
XM_006714757.1 | XP_006714820.1 | 5 - 6 | 1293 | 117 | |
XM_011537567.2 | XP_011535869.1 | 5 - 6 | 1293 | 117 | |
XM_011537568.2 | XP_011535870.1 | 5 - 6 | 1287 | 117 | |
XM_011537569.2 | XP_011535871.1 | 5 - 6 | 1293 | 117 | |
XM_017009136.1 | XP_016864625.1 | 5 - 6 | 1287 | 117 | |
GenBank mRNA | AK074234.1 | - | 5 - 6 | 1281 | 117 |
AK292576.1 | - | 5 - 6 | 1282 | 117 | |
AL833297.1 | - | 4 - 5 | 1111 | 117 | |
BC125106.1 | - | 5 - 6 | 1254 | 117 | |
BC125107.1 | - | 5 - 6 | 1254 | 117 |