Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014175.3 | NP_054894.1 | 2 - 3 | 327 | 108 |
XM_017013362.1 | XP_016868851.1 | 2 - 3 | 338 | 108 | |
GenBank mRNA | AF161494.1 | - | 2 - 3 | 319 | 108 |
AK312832.1 | - | 2 - 3 | 291 | 108 | |
AL136665.1 | - | 2 - 3 | 297 | 108 | |
BC000891.1 | - | 2 - 3 | 327 | 108 | |
CR533531.1 | - | 2 - 3 | 264 | 108 |