Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003750.2 | NP_003741.1 | 8 - 9 | 1372 | 104 |
GenBank mRNA | AK302575.1 | - | 8 - 9 | 1317 | 104 |
AK307610.1 | - | 8 - 9 | 1350 | 104 | |
BC033769.1 | - | 8 - 9 | 1345 | 104 | |
BC054342.1 | - | 8 - 9 | 1331 | 104 | |
BC064926.1 | - | 8 - 9 | 1356 | 104 | |
BC107776.1 | - | 8 - 9 | 1364 | 104 | |
BC114429.1 | - | 8 - 9 | 1280 | 104 | |
D50929.1 | - | 8 - 9 | 1354 | 104 | |
U58046.1 | - | 8 - 9 | 1324 | 104 | |
U78311.1 | - | 8 - 9 | 1339 | 104 |