Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015691.3 | NP_056506.2 | 12 - 13 | 1951 | 137 |
GenBank mRNA | AB033106.1 | - | 10 - 11 | 1574 | 137 |
AK024683.1 | - | 3 - 4 | 761 | 137 | |
AK091936.1 | - | 3 - 4 | 628 | 137 | |
AK292961.1 | - | 11 - 12 | 1833 | 137 | |
AK301129.1 | - | 5 - 6 | 982 | 137 | |
AL353937.1 | - | 9 - 10 | 1442 | 137 | |
BC032447.1 | - | 9 - 10 | 1486 | 137 | |
KC987947.1 | - | 12 - 13 | 2128 | 137 |