Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015691.3 | NP_056506.2 | 19 - 20 | 2960 | 64 |
GenBank mRNA | AB033106.1 | - | 17 - 18 | 2583 | 64 |
AK024683.1 | - | 10 - 11 | 1770 | 64 | |
AK091936.1 | - | 10 - 11 | 1637 | 64 | |
AK292961.1 | - | 18 - 19 | 2842 | 64 | |
AL353937.1 | - | 16 - 17 | 2451 | 64 | |
BC003527.1 | - | 1 - 2 | 137 | 64 | |
BC032447.1 | - | 16 - 17 | 2495 | 64 | |
KC987947.1 | - | 19 - 20 | 3137 | 64 |