Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015691.3 | NP_056506.2 | 4 - 5 | 496 | 75 |
GenBank mRNA | AB033106.1 | - | 2 - 3 | 119 | 75 |
AK292961.1 | - | 3 - 4 | 378 | 75 | |
BU500300.1 | - | 4 - 5 | 704 | 75 | |
KC987947.1 | - | 4 - 5 | 673 | 75 |