Assay Design



Gene Transcripts

Gene Symbol
VPS41
Entrez Gene ID
27072
Gene Name
VPS41, HOPS complex subunit
Gene Aliases
HVPS41, HVSP41, hVps41p
Chromosome Location
Chr.7: 38722974 - 38909200 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014396.3 NP_055211.2 1 - 2 44 296
GenBank mRNA BC044851.1 - 1 - 2 39 296
BX648347.1 - 1 - 2 36 296

View Full Product Details