Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018323.3 | NP_060793.2 | 8 - 9 | 1351 | 168 |
XM_005248174.1 | XP_005248231.1 | 8 - 9 | 1334 | 168 | |
XM_005248175.4 | XP_005248232.1 | 8 - 9 | 1129 | 168 | |
GenBank mRNA | AF411320.1 | - | 8 - 9 | 1218 | 168 |
AK001967.1 | - | 8 - 9 | 1339 | 168 | |
AK023186.1 | - | 1 - 2 | 132 | 168 | |
AK128622.1 | - | 9 - 10 | 1256 | 168 | |
AK296302.1 | - | 8 - 9 | 1041 | 168 | |
AY065990.1 | - | 8 - 9 | 1329 | 168 | |
AY091514.1 | - | 8 - 9 | 1338 | 168 | |
BC051749.1 | - | 8 - 9 | 1351 | 168 |