Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320923.1 | NP_001307852.1 | 21 - 22 | 3056 | 137 |
NM_001321852.1 | NP_001308781.1 | 20 - 21 | 3059 | 137 | |
NM_001321853.1 | NP_001308782.1 | 22 - 23 | 3318 | 137 | |
NM_001321854.1 | NP_001308783.1 | 21 - 22 | 3234 | 137 | |
NM_001321855.1 | NP_001308784.1 | 21 - 22 | 3237 | 137 | |
NM_001321856.1 | NP_001308785.1 | 20 - 21 | 2972 | 137 | |
NM_001321857.1 | NP_001308786.1 | 20 - 21 | 3150 | 137 | |
NM_002227.3 | NP_002218.2 | 20 - 21 | 3153 | 137 | |
XM_017001212.1 | XP_016856701.1 | 20 - 21 | 2997 | 137 | |
GenBank mRNA | AB209057.1 | - | 19 - 20 | 2909 | 137 |
AB219242.1 | - | 18 - 19 | 2800 | 137 | |
AK314257.1 | - | 21 - 22 | 3057 | 137 | |
BC132729.1 | - | 19 - 20 | 2871 | 137 | |
BX647675.1 | - | 6 - 7 | 757 | 137 | |
HQ628631.1 | - | 18 - 19 | 2833 | 137 | |
M64174.1 | - | 19 - 20 | 2878 | 137 |