Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023112.3 | NP_075601.1 | 2 - 3 | 256 | 101 |
XM_005268055.3 | XP_005268112.1 | 3 - 4 | 306 | 101 | |
XM_011537148.2 | XP_011535450.1 | 2 - 3 | 258 | 101 | |
GenBank mRNA | AK025569.1 | - | 2 - 3 | 232 | 101 |
AY177201.1 | - | 1 - 2 | 96 | 101 | |
BC009615.1 | - | 2 - 3 | 256 | 101 | |
BC068058.1 | - | 2 - 3 | 256 | 101 | |
CR457345.1 | - | 1 - 2 | 96 | 101 |