Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020754.3 | NP_065805.2 | 11 - 12 | 2458 | 70 |
XM_006713714.3 | XP_006713777.1 | 11 - 12 | 2482 | 70 | |
XM_017006955.1 | XP_016862444.1 | 10 - 11 | 2397 | 70 | |
GenBank mRNA | AB033030.1 | - | 11 - 12 | 2458 | 70 |
AK293726.1 | - | 11 - 12 | 2055 | 70 | |
BC112163.1 | - | 11 - 12 | 1953 | 70 | |
BC112165.1 | - | 11 - 12 | 1953 | 70 |