Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020754.3 | NP_065805.2 | 1 - 2 | 627 | 63 |
XM_006713714.3 | XP_006713777.1 | 1 - 2 | 711 | 63 | |
XM_017006955.1 | XP_016862444.1 | 1 - 2 | 711 | 63 | |
GenBank mRNA | AB033030.1 | - | 1 - 2 | 627 | 63 |
AK293726.1 | - | 1 - 2 | 224 | 63 | |
BC112163.1 | - | 1 - 2 | 122 | 63 | |
BC112165.1 | - | 1 - 2 | 122 | 63 |