Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020754.3 | NP_065805.2 | 5 - 6 | 1072 | 68 |
XM_006713714.3 | XP_006713777.1 | 5 - 6 | 1156 | 68 | |
XM_017006955.1 | XP_016862444.1 | 4 - 5 | 1011 | 68 | |
GenBank mRNA | AB033030.1 | - | 5 - 6 | 1072 | 68 |
AK293726.1 | - | 5 - 6 | 669 | 68 | |
BC112163.1 | - | 5 - 6 | 567 | 68 | |
BC112165.1 | - | 5 - 6 | 567 | 68 |