Assay Design



Gene Transcripts

Gene Symbol
TRIM31
Entrez Gene ID
11074
Gene Name
tripartite motif containing 31
Gene Aliases
C6orf13, HCG1, HCGI, RNF
Chromosome Location
Chr.6: 30102892 - 30113106 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_134870.1 9 - 10 1465 127
GenBank mRNA AK222729.1 - 9 - 10 1449 127
AK290270.1 - 9 - 10 1452 127
X81006.1 - 3 - 4 875 127

View Full Product Details