Assay Design



Gene Transcripts

Gene Symbol
DTNB
Entrez Gene ID
1838
Gene Name
dystrobrevin beta
Gene Aliases
-
Chromosome Location
Chr.2: 25377220 - 25673647 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256303.1 NP_001243232.1 6 - 7 869 76
NM_001256304.1 NP_001243233.1 5 - 6 606 76
NM_001256308.2 NP_001243237.1 5 - 6 740 76
NM_001320933.1 NP_001307862.1 6 - 7 869 76
NM_001320934.1 NP_001307863.1 6 - 7 869 76
NM_001320936.1 NP_001307865.1 6 - 7 869 76
NM_001320937.1 NP_001307866.1 6 - 7 869 76
NM_021907.4 NP_068707.1 6 - 7 869 76
NM_033147.3 NP_149159.2 6 - 7 869 76
NM_033148.3 NP_149160.1 6 - 7 869 76
NM_183360.2 NP_899204.1 6 - 7 869 76
NM_183361.2 NP_899205.1 6 - 7 869 76
XM_005264182.1 XP_005264239.1 6 - 7 786 76
XM_011532671.1 XP_011530973.1 6 - 7 786 76
XM_011532672.2 XP_011530974.1 6 - 7 786 76
XM_011532673.1 XP_011530975.1 6 - 7 786 76
XM_011532674.1 XP_011530976.1 6 - 7 786 76
XM_011532675.1 XP_011530977.1 6 - 7 786 76
XM_011532676.1 XP_011530978.1 6 - 7 786 76
XM_011532677.2 XP_011530979.1 5 - 6 1242 76
XM_011532679.1 XP_011530981.1 6 - 7 786 76
XM_011532680.2 XP_011530982.1 5 - 6 707 76
XM_011532681.2 XP_011530983.1 5 - 6 700 76
XM_017003529.1 XP_016859018.1 6 - 7 786 76
XM_017003530.1 XP_016859019.1 6 - 7 786 76
XM_017003531.1 XP_016859020.1 6 - 7 786 76
XM_017003532.1 XP_016859021.1 6 - 7 786 76
XM_017003533.1 XP_016859022.1 6 - 7 786 76
XM_017003534.1 XP_016859023.1 6 - 7 786 76
XM_017003535.1 XP_016859024.1 6 - 7 786 76
XM_017003536.1 XP_016859025.1 6 - 7 786 76
XM_017003537.1 XP_016859026.1 2 - 3 1139 76
XM_017003538.1 XP_016859027.1 5 - 6 632 76
XM_017003539.1 XP_016859028.1 5 - 6 643 76
XM_017003540.1 XP_016859029.1 5 - 6 700 76
XM_017003541.1 XP_016859030.1 5 - 6 684 76
GenBank mRNA AF022728.1 - 6 - 7 767 76
AI608704.1 - 3 - 4 76
AK294357.1 - 5 - 6 550 76
AK300019.1 - 6 - 7 746 76
AK301386.1 - 5 - 6 742 76
AL555818.3 - 6 - 7 783 76
BC016655.1 - 6 - 7 762 76
BC049366.1 - 6 - 7 860 76
BT009805.1 - 5 - 6 606 76
BU846095.1 - 4 - 5 547 76
DQ160290.1 - 5 - 6 606 76
Y12712.1 - 6 - 7 789 76
Y15718.1 - 5 - 6 606 76
Y15722.1 - 5 - 6 606 76

View Full Product Details