Assay Design



Gene Transcripts

Gene Symbol
ZFPL1
Entrez Gene ID
7542
Gene Name
zinc finger protein like 1
Gene Aliases
D11S750, MCG4
Chromosome Location
Chr.11: 65084222 - 65088402 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006782.3 NP_006773.2 2 - 3 268 59
GenBank mRNA AF001891.1 - 2 - 3 250 59
AF030291.1 - 2 - 3 255 59
AF043611.1 - 2 - 3 132 59
AK291964.1 - 2 - 3 249 59
AK295279.1 - 2 - 3 231 59
AK308740.1 - 2 - 3 239 59
AK316044.1 - 2 - 3 248 59
AU279924.1 - 2 - 3 268 59
BC012530.2 - 2 - 3 203 59
BC012814.2 - 2 - 3 203 59

View Full Product Details