Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005483.2 | NP_005474.2 | 5 - 6 | 1348 | 92 |
XM_011527605.2 | XP_011525907.1 | 5 - 6 | 1415 | 92 | |
XM_011527606.2 | XP_011525908.1 | 5 - 6 | 1415 | 92 | |
XM_011527607.2 | XP_011525909.1 | 4 - 5 | 1358 | 92 | |
GenBank mRNA | BC067093.1 | - | 5 - 6 | 1349 | 92 |
U20979.1 | - | 5 - 6 | 1234 | 92 |