Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005483.2 | NP_005474.2 | 6 - 7 | 1411 | 99 |
XM_011527605.2 | XP_011525907.1 | 6 - 7 | 1478 | 99 | |
XM_011527606.2 | XP_011525908.1 | 6 - 7 | 1478 | 99 | |
XM_011527607.2 | XP_011525909.1 | 5 - 6 | 1421 | 99 | |
GenBank mRNA | BC067093.1 | - | 6 - 7 | 1412 | 99 |
U20979.1 | - | 6 - 7 | 1297 | 99 |