Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005483.2 | NP_005474.2 | 8 - 9 | 1705 | 103 |
XM_011527605.2 | XP_011525907.1 | 8 - 9 | 1772 | 103 | |
XM_011527606.2 | XP_011525908.1 | 8 - 9 | 1772 | 103 | |
XM_011527607.2 | XP_011525909.1 | 7 - 8 | 1715 | 103 | |
GenBank mRNA | BC067093.1 | - | 8 - 9 | 1706 | 103 |
U20979.1 | - | 8 - 9 | 1591 | 103 |