Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016195.3 | NP_057279.2 | 15 - 16 | 2094 | 112 |
GenBank mRNA | AK074336.1 | - | 15 - 16 | 2044 | 113 |
AK290439.1 | - | 15 - 16 | 2074 | 112 | |
AL117496.1 | - | 15 - 16 | 2049 | 112 | |
AL137392.1 | - | 2 - 3 | 681 | 112 | |
AY282406.1 | - | 15 - 16 | 2071 | 112 | |
BC042130.1 | - | 15 - 16 | 2046 | 112 | |
BC058913.1 | - | 15 - 16 | 2050 | 112 |