Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284259.1 | NP_001271188.1 | 20 - 21 | 3864 | 146 |
NM_016195.3 | NP_057279.2 | 20 - 21 | 3744 | 146 | |
GenBank mRNA | AB033337.1 | - | 20 - 21 | 3817 | 146 |
AK024959.1 | - | 3 - 4 | 1439 | 146 | |
AL117496.1 | - | 20 - 21 | 3699 | 146 | |
AL137392.1 | - | 7 - 8 | 2331 | 146 | |
AY282407.1 | - | 5 - 6 | 1661 | 146 | |
AY739715.1 | - | 1 - 2 | 457 | 146 | |
U93121.1 | - | 1 - 2 | 572 | 146 |