Assay Design



Gene Transcripts

Gene Symbol
AKIP1
Entrez Gene ID
56672
Gene Name
A-kinase interacting protein 1
Gene Aliases
BCA3, C11orf17
Chromosome Location
Chr.11: 8911117 - 8920079 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206647.1 NP_001193576.1 3 - 4 376 105
NM_020642.3 NP_065693.2 3 - 4 376 105
NR_045417.1 2 - 3 558 105
XM_017018011.1 XP_016873500.1 3 - 4 396 105
GenBank mRNA AF493783.1 - 3 - 4 376 105
AF493784.1 - 3 - 4 376 105
AF512007.1 - 2 - 3 300 105
AK294549.1 - 2 - 3 537 105
BC030996.2 - 2 - 3 306 105

View Full Product Details