Assay Design



Gene Transcripts

Gene Symbol
HSF1
Entrez Gene ID
3297
Gene Name
heat shock transcription factor 1
Gene Aliases
HSTF1
Chromosome Location
Chr.8: 144291588 - 144314722 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005526.2 NP_005517.1 11 - 12 1482 96
XM_005272315.2 XP_005272372.1 12 - 13 1566 96
XM_005272316.2 XP_005272373.1 12 - 13 1566 96
XM_011517004.2 XP_011515306.1 9 - 10 1091 96
XM_011517006.2 XP_011515308.1 7 - 8 873 96
XM_017013377.1 XP_016868866.1 11 - 12 1482 96
GenBank mRNA AF299094.1 - 3 - 4 202 96
AK125467.1 - 8 - 9 2784 96
AK222497.1 - 11 - 12 1466 96
AK290975.1 - 11 - 12 1462 96
AK300205.1 - 8 - 9 964 96
BC014638.1 - 11 - 12 1453 96
BT007351.1 - 11 - 12 1312 96
M64673.1 - 11 - 12 1472 96

View Full Product Details