Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005526.2 | NP_005517.1 | 6 - 7 | 802 | 75 |
XM_005272315.2 | XP_005272372.1 | 6 - 7 | 802 | 75 | |
XM_005272316.2 | XP_005272373.1 | 6 - 7 | 802 | 75 | |
XM_005272317.1 | XP_005272374.1 | 6 - 7 | 802 | 75 | |
XM_011517004.2 | XP_011515306.1 | 3 - 4 | 327 | 75 | |
XM_011517006.2 | XP_011515308.1 | 1 - 2 | 68 | 75 | |
XM_017013377.1 | XP_016868866.1 | 6 - 7 | 802 | 75 | |
GenBank mRNA | AK054762.1 | - | 2 - 3 | 919 | 75 |
AK125467.1 | - | 3 - 4 | 2104 | 75 | |
AK290975.1 | - | 6 - 7 | 782 | 75 | |
AK300205.1 | - | 3 - 4 | 284 | 75 | |
BC014638.1 | - | 6 - 7 | 773 | 75 | |
BT007351.1 | - | 6 - 7 | 632 | 75 | |
M64673.1 | - | 6 - 7 | 792 | 75 |