Assay Design



Gene Transcripts

Gene Symbol
MFAP2
Entrez Gene ID
4237
Gene Name
microfibrillar associated protein 2
Gene Aliases
MAGP, MAGP-1, MAGP1
Chromosome Location
Chr.1: 16974502 - 16981649 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135247.1 NP_001128719.1 4 - 5 267 78
NM_001135248.1 NP_001128720.1 4 - 5 286 78
NM_002403.3 NP_002394.1 4 - 5 289 78
NM_017459.2 NP_059453.1 4 - 5 270 78
XM_011541476.2 XP_011539778.1 4 - 5 896 78
GenBank mRNA AK222751.1 - 4 - 5 248 78
BC015039.1 - 4 - 5 289 78
CR407678.1 - 3 - 4 156 78
DN996554.1 - 4 - 5 239 78
U19718.1 - 3 - 4 197 78

View Full Product Details