Assay Design



Gene Transcripts

Gene Symbol
NRXN3
Entrez Gene ID
9369
Gene Name
neurexin 3
Gene Aliases
C14orf60
Chromosome Location
Chr.14: 77979904 - 79868290 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004796.5 NP_004787.2 10 - 11 2360 67
NR_073547.1 12 - 13 3941 67
XM_005268218.2 XP_005268275.1 15 - 16 4010 67
XM_006720322.2 XP_006720385.1 14 - 15 3998 67
XM_006720323.2 XP_006720386.1 13 - 14 3974 67
XM_011537363.1 XP_011535665.1 15 - 16 4010 67
XM_011537364.2 XP_011535666.1 16 - 17 3972 67
XM_011537365.2 XP_011535667.1 15 - 16 3798 67
XM_011537366.1 XP_011535668.1 14 - 15 3986 67
XM_011537367.1 XP_011535669.1 15 - 16 4010 67
XM_011537368.1 XP_011535670.1 15 - 16 4010 67
XM_011537369.1 XP_011535671.1 15 - 16 4010 67
XM_011537370.1 XP_011535672.1 14 - 15 3986 67
XM_011537371.1 XP_011535673.1 15 - 16 4010 67
XM_011537372.1 XP_011535674.1 15 - 16 4010 67
XM_011537373.1 XP_011535675.1 15 - 16 4010 67
XM_011537377.2 XP_011535679.1 11 - 12 2302 67
XM_017021790.1 XP_016877279.1 16 - 17 4110 67
XM_017021791.1 XP_016877280.1 16 - 17 3893 67
XM_017021792.1 XP_016877281.1 14 - 15 3793 67
XM_017021793.1 XP_016877282.1 14 - 15 3983 67
XM_017021794.1 XP_016877283.1 13 - 14 3974 67
XM_017021795.1 XP_016877284.1 13 - 14 3769 67
XM_017021796.1 XP_016877285.1 13 - 14 3959 67
XM_017021797.1 XP_016877286.1 12 - 13 3950 67
XM_017021798.1 XP_016877287.1 12 - 13 3947 67
XM_017021799.1 XP_016877288.1 15 - 16 4010 67
XM_017021800.1 XP_016877289.1 15 - 16 4010 67
XM_017021801.1 XP_016877290.1 14 - 15 3983 67
XM_017021802.1 XP_016877291.1 13 - 14 4004 67
XM_017021803.1 XP_016877292.1 14 - 15 3989 67
XM_017021804.1 XP_016877293.1 14 - 15 3983 67
XM_017021805.1 XP_016877294.1 13 - 14 3974 67
XM_017021806.1 XP_016877295.1 13 - 14 3959 67
XM_017021807.1 XP_016877296.1 15 - 16 4010 67
GenBank mRNA AB018286.1 - 10 - 11 2341 67
AJ316284.2 - 12 - 13 3729 67
AK310235.1 - 9 - 10 2286 67
BC152457.1 - 10 - 11 2341 67

View Full Product Details