Assay Design



Gene Transcripts

Gene Symbol
NRXN3
Entrez Gene ID
9369
Gene Name
neurexin 3
Gene Aliases
C14orf60
Chromosome Location
Chr.14: 77979904 - 79868290 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004796.5 NP_004787.2 6 - 7 1637 86
XM_005268218.2 XP_005268275.1 11 - 12 3287 86
XM_006720322.2 XP_006720385.1 10 - 11 3275 86
XM_006720323.2 XP_006720386.1 9 - 10 3251 86
XM_011537363.1 XP_011535665.1 11 - 12 3287 86
XM_011537364.2 XP_011535666.1 12 - 13 3249 86
XM_011537365.2 XP_011535667.1 11 - 12 3075 86
XM_011537366.1 XP_011535668.1 10 - 11 3263 86
XM_011537367.1 XP_011535669.1 11 - 12 3287 86
XM_011537368.1 XP_011535670.1 11 - 12 3287 86
XM_011537369.1 XP_011535671.1 11 - 12 3287 86
XM_011537370.1 XP_011535672.1 10 - 11 3263 86
XM_011537371.1 XP_011535673.1 11 - 12 3287 86
XM_011537372.1 XP_011535674.1 11 - 12 3287 86
XM_011537373.1 XP_011535675.1 11 - 12 3287 86
XM_011537377.2 XP_011535679.1 7 - 8 1579 86
XM_017021790.1 XP_016877279.1 12 - 13 3387 86
XM_017021791.1 XP_016877280.1 12 - 13 3170 86
XM_017021792.1 XP_016877281.1 10 - 11 3070 86
XM_017021794.1 XP_016877283.1 9 - 10 3251 86
XM_017021795.1 XP_016877284.1 9 - 10 3046 86
XM_017021797.1 XP_016877286.1 8 - 9 3227 86
XM_017021799.1 XP_016877288.1 11 - 12 3287 86
XM_017021800.1 XP_016877289.1 11 - 12 3287 86
XM_017021802.1 XP_016877291.1 9 - 10 3281 86
XM_017021803.1 XP_016877292.1 10 - 11 3266 86
XM_017021805.1 XP_016877294.1 9 - 10 3251 86
XM_017021807.1 XP_016877296.1 11 - 12 3287 86
GenBank mRNA AB018286.1 - 6 - 7 1618 86
BC152457.1 - 6 - 7 1618 86

View Full Product Details