Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022662.3 | NP_073153.1 | 16 - 17 | 2096 | 85 |
XM_011511633.2 | XP_011509935.1 | 16 - 17 | 2128 | 85 | |
XM_011511634.2 | XP_011509936.1 | 16 - 17 | 2243 | 85 | |
XM_011511636.2 | XP_011509938.1 | 16 - 17 | 2128 | 85 | |
XM_011511637.2 | XP_011509939.1 | 16 - 17 | 2128 | 85 | |
XM_017004710.1 | XP_016860199.1 | 16 - 17 | 2128 | 85 | |
XM_017004711.1 | XP_016860200.1 | 16 - 17 | 2128 | 85 | |
XM_017004712.1 | XP_016860201.1 | 16 - 17 | 2243 | 85 | |
XM_017004713.1 | XP_016860202.1 | 15 - 16 | 2191 | 85 | |
XM_017004714.1 | XP_016860203.1 | 15 - 16 | 2139 | 85 | |
XM_017004715.1 | XP_016860204.1 | 16 - 17 | 2128 | 85 | |
GenBank mRNA | AJ278357.1 | - | 16 - 17 | 2111 | 85 |
AL832013.1 | - | 5 - 6 | 452 | 85 | |
BC104902.1 | - | 16 - 17 | 1898 | 85 | |
BC104904.1 | - | 16 - 17 | 1898 | 85 |