Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022662.3 | NP_073153.1 | 22 - 23 | 2954 | 147 |
XM_011511633.2 | XP_011509935.1 | 22 - 23 | 2986 | 147 | |
XM_011511634.2 | XP_011509936.1 | 22 - 23 | 3101 | 147 | |
XM_011511636.2 | XP_011509938.1 | 22 - 23 | 2986 | 147 | |
XM_011511637.2 | XP_011509939.1 | 22 - 23 | 2986 | 147 | |
XM_017004710.1 | XP_016860199.1 | 22 - 23 | 2986 | 147 | |
XM_017004711.1 | XP_016860200.1 | 22 - 23 | 2986 | 147 | |
XM_017004712.1 | XP_016860201.1 | 22 - 23 | 3101 | 147 | |
XM_017004713.1 | XP_016860202.1 | 21 - 22 | 3049 | 147 | |
XM_017004714.1 | XP_016860203.1 | 21 - 22 | 2997 | 147 | |
XM_017004715.1 | XP_016860204.1 | 22 - 23 | 2986 | 147 | |
GenBank mRNA | AJ278357.1 | - | 22 - 23 | 2969 | 147 |
AK023807.1 | - | 9 - 10 | 1180 | 147 | |
AL832013.1 | - | 11 - 12 | 1310 | 147 | |
BC104902.1 | - | 22 - 23 | 2756 | 147 | |
BC104904.1 | - | 22 - 23 | 2756 | 147 |