Assay Design
Gene Transcripts
- Gene Symbol
- ANAPC1
- Entrez Gene ID
-
64682
- Gene Name
- anaphase promoting complex subunit 1
- Gene Aliases
-
APC1, MCPR, TSG24
- Chromosome Location
-
Chr.2: 111766147 - 111884196 on Build GRCh38
- Gene Symbol
- ANAPC1P1
- Entrez Gene ID
-
100286979
- Gene Name
- anaphase promoting complex subunit 1 pseudogene 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 86861825 - 86895042 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_037931.1 |
|
2 - 3 |
338 |
107 |
GenBank mRNA |
AK310642.1 |
- |
2 - 3 |
338 |
107 |
View Full Product Details