Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022662.3 | NP_073153.1 | 4 - 5 | 670 | 160 |
XM_011511633.2 | XP_011509935.1 | 4 - 5 | 702 | 160 | |
XM_011511634.2 | XP_011509936.1 | 4 - 5 | 817 | 160 | |
XM_011511636.2 | XP_011509938.1 | 4 - 5 | 702 | 160 | |
XM_011511637.2 | XP_011509939.1 | 4 - 5 | 702 | 160 | |
XM_017004710.1 | XP_016860199.1 | 4 - 5 | 702 | 160 | |
XM_017004711.1 | XP_016860200.1 | 4 - 5 | 702 | 160 | |
XM_017004712.1 | XP_016860201.1 | 4 - 5 | 817 | 160 | |
XM_017004715.1 | XP_016860204.1 | 4 - 5 | 702 | 160 | |
GenBank mRNA | AJ278357.1 | - | 4 - 5 | 685 | 160 |
BC104902.1 | - | 4 - 5 | 472 | 160 | |
BC104904.1 | - | 4 - 5 | 472 | 160 |