Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008222.2 | NP_001008223.1 | 5 - 6 | 1083 | 124 |
NM_016032.3 | NP_057116.2 | 6 - 7 | 1063 | 124 | |
GenBank mRNA | AB209807.1 | - | 5 - 6 | 1002 | 124 |
AF151847.1 | - | 6 - 7 | 951 | 124 | |
AK001112.1 | - | 6 - 7 | 1013 | 124 | |
AK001424.1 | - | 3 - 4 | 397 | 124 | |
AK001524.1 | - | 5 - 6 | 814 | 124 | |
AK096967.1 | - | 6 - 7 | 714 | 124 | |
AL161962.1 | - | 6 - 7 | 940 | 124 | |
AY358558.1 | - | 6 - 7 | 995 | 124 | |
AY952881.8 | - | 6 - 7 | 1035 | 124 | |
BC000035.2 | - | 5 - 6 | 957 | 124 | |
BC003128.1 | - | 6 - 7 | 1001 | 124 | |
BC006200.2 | - | 6 - 7 | 997 | 124 | |
BC012826.2 | - | 6 - 7 | 997 | 124 |