Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004500.3 | NP_004491.2 | 2 - 3 | 208 | 86 |
NM_031314.2 | NP_112604.2 | 2 - 3 | 208 | 86 | |
GenBank mRNA | AB209527.1 | - | 2 - 3 | 142 | 86 |
AK223517.1 | - | 2 - 3 | 148 | 86 | |
AK225032.1 | - | 2 - 3 | 171 | 86 | |
AK298906.1 | - | 2 - 3 | 155 | 86 | |
AK299923.1 | - | 2 - 3 | 153 | 86 | |
AK312338.1 | - | 2 - 3 | 153 | 86 | |
BC003394.1 | - | 2 - 3 | 139 | 86 | |
BC008364.1 | - | 2 - 3 | 146 | 86 | |
BC092433.1 | - | 2 - 3 | 156 | 86 | |
BX247961.1 | - | 2 - 3 | 139 | 86 | |
M16342.1 | - | 2 - 3 | 86 | 86 | |
M29063.1 | - | 2 - 3 | 143 | 86 |