Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004227.3 | NP_004218.1 | 11 - 12 | 1076 | 75 |
GenBank mRNA | AJ005197.1 | - | 11 - 12 | 1047 | 75 |
AJ223957.1 | - | 11 - 12 | 997 | 75 | |
AK295163.1 | - | 6 - 7 | 868 | 75 | |
AK313728.1 | - | 12 - 13 | 1092 | 75 | |
BC008191.1 | - | 4 - 5 | 315 | 75 | |
BC028717.1 | - | 11 - 12 | 1112 | 75 |