Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130158.1 | NP_001123630.1 | 18 - 19 | 2239 | 156 |
NM_001161819.1 | NP_001155291.1 | 18 - 19 | 2077 | 156 | |
NM_012223.3 | NP_036355.2 | 18 - 19 | 2239 | 156 | |
XM_005246572.1 | XP_005246629.1 | 18 - 19 | 2103 | 156 | |
XM_017004156.1 | XP_016859645.1 | 18 - 19 | 2212 | 156 | |
XM_017004157.1 | XP_016859646.1 | 18 - 19 | 2212 | 156 | |
XM_017004158.1 | XP_016859647.1 | 16 - 17 | 1828 | 156 | |
GenBank mRNA | AB290176.1 | - | 18 - 19 | 2256 | 156 |
AJ001381.1 | - | 1 - 2 | 57 | 156 | |
AK000160.1 | - | 14 - 15 | 1573 | 156 | |
BC053558.1 | - | 18 - 19 | 2077 | 156 |