Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014641.2 | NP_055456.2 | 14 - 15 | 6538 | 64 |
XM_005249492.2 | XP_005249549.1 | 14 - 15 | 6442 | 64 | |
XM_005249493.3 | XP_005249550.1 | 14 - 15 | 6439 | 64 | |
XM_005249494.4 | XP_005249551.1 | 14 - 15 | 6841 | 64 | |
XM_005249497.3 | XP_005249554.1 | 14 - 15 | 5872 | 64 | |
XM_005249498.3 | XP_005249555.1 | 13 - 14 | 5650 | 64 | |
XM_011515001.2 | XP_011513303.1 | 14 - 15 | 6269 | 64 | |
XM_011515003.2 | XP_011513305.1 | 14 - 15 | 6124 | 64 | |
XM_011515004.2 | XP_011513306.1 | 15 - 16 | 6153 | 64 | |
XM_017011519.1 | XP_016867008.1 | 14 - 15 | 6221 | 64 | |
XM_017011520.1 | XP_016867009.1 | 14 - 15 | 6137 | 64 | |
XM_017011521.1 | XP_016867010.1 | 14 - 15 | 5570 | 64 | |
XM_017011522.1 | XP_016867011.1 | 13 - 14 | 5647 | 64 | |
GenBank mRNA | BC110645.1 | - | 9 - 10 | 2195 | 64 |
BC131491.1 | - | 10 - 11 | 3509 | 64 | |
BC152556.1 | - | 14 - 15 | 5250 | 64 | |
CR749828.1 | - | 10 - 11 | 4170 | 64 | |
D79992.1 | - | 13 - 14 | 6111 | 64 |