Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014641.2 | NP_055456.2 | 4 - 5 | 1029 | 111 |
XM_005249492.2 | XP_005249549.1 | 4 - 5 | 933 | 111 | |
XM_005249493.3 | XP_005249550.1 | 4 - 5 | 933 | 111 | |
XM_005249494.4 | XP_005249551.1 | 4 - 5 | 1332 | 111 | |
XM_005249497.3 | XP_005249554.1 | 4 - 5 | 933 | 111 | |
XM_005249498.3 | XP_005249555.1 | 4 - 5 | 933 | 111 | |
XM_011515001.2 | XP_011513303.1 | 4 - 5 | 760 | 111 | |
XM_011515003.2 | XP_011513305.1 | 4 - 5 | 615 | 111 | |
XM_011515004.2 | XP_011513306.1 | 5 - 6 | 644 | 111 | |
XM_017011519.1 | XP_016867008.1 | 4 - 5 | 712 | 111 | |
XM_017011520.1 | XP_016867009.1 | 4 - 5 | 631 | 111 | |
XM_017011521.1 | XP_016867010.1 | 4 - 5 | 631 | 111 | |
XM_017011522.1 | XP_016867011.1 | 4 - 5 | 933 | 111 | |
GenBank mRNA | AK302434.1 | - | 5 - 6 | 635 | 111 |
BC131491.1 | - | 4 - 5 | 1029 | 111 | |
BC152556.1 | - | 3 - 4 | 602 | 111 | |
CR749828.1 | - | 4 - 5 | 707 | 111 | |
D79992.1 | - | 3 - 4 | 602 | 111 | |
EF177820.1 | - | 4 - 5 | 657 | 111 | |
EF177821.1 | - | 4 - 5 | 731 | 111 | |
EF177822.1 | - | 5 - 6 | 688 | 111 |