Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014641.2 | NP_055456.2 | 9 - 10 | 3523 | 81 |
XM_005249492.2 | XP_005249549.1 | 9 - 10 | 3427 | 81 | |
XM_005249493.3 | XP_005249550.1 | 9 - 10 | 3424 | 81 | |
XM_005249494.4 | XP_005249551.1 | 9 - 10 | 3826 | 81 | |
XM_005249497.3 | XP_005249554.1 | 9 - 10 | 2857 | 81 | |
XM_005249498.3 | XP_005249555.1 | 8 - 9 | 2635 | 81 | |
XM_011515001.2 | XP_011513303.1 | 9 - 10 | 3254 | 81 | |
XM_011515003.2 | XP_011513305.1 | 9 - 10 | 3109 | 81 | |
XM_011515004.2 | XP_011513306.1 | 10 - 11 | 3138 | 81 | |
XM_017011519.1 | XP_016867008.1 | 9 - 10 | 3206 | 81 | |
XM_017011520.1 | XP_016867009.1 | 9 - 10 | 3122 | 81 | |
XM_017011521.1 | XP_016867010.1 | 9 - 10 | 2555 | 81 | |
XM_017011522.1 | XP_016867011.1 | 8 - 9 | 2632 | 81 | |
GenBank mRNA | AK302434.1 | - | 9 - 10 | 2334 | 81 |