Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001826.2 | NP_001817.1 | 1 - 2 | 157 | 100 |
GenBank mRNA | AF274941.1 | - | 1 - 1 | 79 | 100 |
AF279897.1 | - | 1 - 2 | 79 | 100 | |
AK311769.1 | - | 1 - 2 | 118 | 100 | |
BC007751.2 | - | 1 - 2 | 100 | 100 | |
BC015629.2 | - | 1 - 2 | 79 | 100 | |
BC070319.1 | - | 1 - 2 | 118 | 100 | |
BC070320.1 | - | 1 - 2 | 118 | 100 | |
BQ883992.1 | - | 1 - 2 | 96 | 100 | |
CN347951.1 | - | 1 - 2 | 159 | 100 |