Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190737.1 | NP_001177666.1 | 3 - 4 | 112 | |
NM_001190738.1 | NP_001177667.1 | 3 - 4 | 112 | ||
NM_001282787.1 | NP_001269716.1 | 2 - 3 | 112 | ||
NM_005596.3 | NP_005587.2 | 3 - 4 | 112 | ||
XM_005251467.2 | XP_005251524.1 | 3 - 4 | 112 | ||
XM_005251468.4 | XP_005251525.1 | 3 - 4 | 112 | ||
XM_005251469.2 | XP_005251526.1 | 3 - 4 | 112 | ||
XM_005251470.2 | XP_005251527.1 | 3 - 4 | 112 | ||
XM_005251471.3 | XP_005251528.1 | 2 - 3 | 112 | ||
XM_006716773.2 | XP_006716836.1 | 3 - 4 | 112 | ||
XM_006716774.2 | XP_006716837.1 | 3 - 4 | 112 | ||
XM_006716775.2 | XP_006716838.1 | 3 - 4 | 112 | ||
XM_017014739.1 | XP_016870228.1 | 3 - 4 | 112 | ||
XM_017014740.1 | XP_016870229.1 | 3 - 4 | 112 | ||
XM_017014741.1 | XP_016870230.1 | 3 - 4 | 112 | ||
XM_017014742.1 | XP_016870231.1 | 2 - 3 | 112 | ||
GenBank mRNA | AK290603.1 | - | 3 - 4 | 112 | |
AK299605.1 | - | 3 - 4 | 1013 | 112 | |
BC001283.1 | - | 3 - 4 | 112 | ||
BP372854.1 | - | 2 - 3 | 112 | ||
BT007266.1 | - | 3 - 4 | 112 | ||
BX648416.1 | - | 2 - 3 | 112 | ||
BX648845.1 | - | 3 - 4 | 112 | ||
KU178232.1 | - | 3 - 4 | 112 | ||
KU178233.1 | - | 4 - 5 | 112 | ||
U07810.1 | - | 2 - 3 | 112 | ||
U85193.1 | - | 3 - 4 | 112 |