Assay Design
Gene Transcripts
- Gene Symbol
- NFIB
- Entrez Gene ID
-
4781
- Gene Name
- nuclear factor I B
- Gene Aliases
-
CTF, HMGIC/NFIB, NF-I/B, NF1-B, NFI-B, NFI-RED, NFIB2, NFIB3
- Chromosome Location
-
Chr.9: 14081843 - 14398983 on Build GRCh38
View Full Product Details