Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013399.1 | NP_001013417.1 | 3 - 4 | 290 | 153 |
NM_005190.3 | NP_005181.2 | 3 - 4 | 512 | 153 | |
XM_006715594.1 | XP_006715657.1 | 3 - 4 | 671 | 153 | |
XM_011536232.2 | XP_011534534.1 | 3 - 4 | 671 | 153 | |
XM_017011436.1 | XP_016866925.1 | 3 - 4 | 671 | 153 | |
GenBank mRNA | AB125137.1 | - | 3 - 4 | 299 | 153 |
AK222768.1 | - | 3 - 4 | 326 | 153 | |
AK223247.1 | - | 3 - 4 | 313 | 153 | |
AK298558.1 | - | 2 - 3 | 251 | 153 | |
BC010135.2 | - | 3 - 4 | 345 | 153 | |
BC026272.1 | - | 3 - 4 | 326 | 153 | |
BC041123.1 | - | 3 - 4 | 495 | 153 | |
BC056153.1 | - | 3 - 4 | 334 | 153 | |
BM788717.1 | - | 3 - 4 | 290 | 153 | |
M74091.1 | - | 3 - 4 | 313 | 153 |