Assay Design



Gene Transcripts

Gene Symbol
MDGA1
Entrez Gene ID
266727
Gene Name
MAM domain containing glycosylphosphatidylinositol anchor 1
Gene Aliases
GPIM, MAMDC3
Chromosome Location
Chr.6: 37632508 - 37697990 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153487.3 NP_705691.1 3 - 4 1559 60
XM_006715056.3 XP_006715119.1 3 - 4 503 60
XM_017010734.1 XP_016866223.1 3 - 4 503 60
GenBank mRNA AF478693.1 - 3 - 4 1559 60
AK091149.1 - 2 - 3 196 60

View Full Product Details